missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzonitrile, 95%

Product Code. 11467096
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11467096

Brand: Thermo Scientific Alfa Aesar B25416.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17420-30-3 | C7H5N3O2 | 163.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17420-30-3
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00007362
InChI Key MGCGMYPNXAFGFA-UHFFFAOYSA-N
Synonym 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j
PubChem CID 28532
IUPAC Name 2-amino-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)N

Specifications

Melting Point 207°C to 210°C
Quantity 25 g
Formula Weight 163.14
Percent Purity 95%
Chemical Name or Material 2-Amino-5-nitrobenzonitrile
compliance-icons
Product Identifier
  • 2-Amino-5-nitrobenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : 2-Amino-5-nitrobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.