missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals

Product Code. 11489886
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.

Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol, benzene, acetone, and diethyl ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

Melting Point ∼200°C (decomposition)
Quantity 100 g
Beilstein 972974
Solubility Information Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol,benzene,acetone,and diethyl ether.
Formula Weight 154.13
Percent Purity 96%
Chemical Name or Material 2-Amino-5-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'