missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiazole, 97%, Thermo Scientific Chemicals

Product Code. 11449257
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3H3N3O2S
Molecular Weight (g/mol) 145.14
MDL Number MFCD00005326
InChI Key MIHADVKEHAFNPG-UHFFFAOYSA-N
Synonym 2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
PubChem CID 8486
ChEBI CHEBI:82386
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES NC1=NC=C(S1)[N+]([O-])=O

Specifications

Melting Point ∼202°C (decomposition)
Quantity 25 g
Beilstein 126797
Sensitivity Light sensitive
Merck Index 14,457
Formula Weight 145.14
Percent Purity 97%
Chemical Name or Material 2-Amino-5-nitrothiazole
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.