missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromobenzoic acid, 97+%, Thermo Scientific Chemicals

Product Code. 15424787
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20776-48-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD03618455
InChI Key BNQPROAXWQCNKO-UHFFFAOYSA-N
Synonym 6-bromoanthranilic acid, 3-bromo-2-carboxyaniline, benzoic acid, 2-amino-6-bromo, buttpark 49\07-50, 2-amino-6-bromo-benzoic acid, ksc201s8r
PubChem CID 10560649
IUPAC Name 2-amino-6-bromobenzoic acid
SMILES NC1=C(C(O)=O)C(Br)=CC=C1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 216.03
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'