Learn More
2-Amino-6-bromobenzothiazole, 98%, Thermo Scientific Chemicals
£23.20 - £332.00
Chemical Identifiers
CAS | 15864-32-1 |
---|---|
Molecular Formula | C7H5BrN2S |
Molecular Weight (g/mol) | 229.10 |
MDL Number | MFCD00152229 |
InChI Key | VZEBSJIOUMDNLY-UHFFFAOYSA-N |
Synonym | 2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304 |
PubChem CID | 85149 |
IUPAC Name | 6-bromo-1,3-benzothiazol-2-amine |
SMILES | NC1=NC2=CC=C(Br)C=C2S1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11385199
|
Thermo Scientific Alfa Aesar
A18554.03 |
1 g |
£23.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395199
|
Thermo Scientific Alfa Aesar
A18554.06 |
5 g |
£84.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11305209
|
Thermo Scientific Alfa Aesar
A18554.14 |
25 g |
£332.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15864-32-1 | |
229.10 | |
VZEBSJIOUMDNLY-UHFFFAOYSA-N | |
85149 | |
NC1=NC2=CC=C(Br)C=C2S1 |
C7H5BrN2S | |
MFCD00152229 | |
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304 | |
6-bromo-1,3-benzothiazol-2-amine |
Specifications
15864-32-1 | |
C7H5BrN2S | |
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304 | |
NC1=NC2=CC=C(Br)C=C2S1 | |
229.10 | |
229.1 | |
2-Amino-6-bromobenzothiazole |
Odorless | |
MFCD00152229 | |
VZEBSJIOUMDNLY-UHFFFAOYSA-N | |
6-bromo-1,3-benzothiazol-2-amine | |
85149 | |
98% |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only