Learn More
2-Amino-6-bromopyridine, 98%, Thermo Scientific Chemicals
£62.71 - £603.00
Chemical Identifiers
CAS | 19798-81-3 |
---|---|
Molecular Formula | C5H6BrN2 |
Molecular Weight (g/mol) | 174.02 |
MDL Number | MFCD00137843 |
InChI Key | BKLJUYPLUWUEOQ-UHFFFAOYSA-O |
Synonym | 2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 |
PubChem CID | 300809 |
IUPAC Name | 6-bromopyridin-2-amine |
SMILES | NC1=CC=CC(Br)=[NH+]1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15720869
|
Thermo Scientific Acros
376870050 |
5 g | Glass bottle |
£62.71
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10082282
|
Thermo Scientific Acros
376870250 |
25 g | Glass bottle |
£204.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926331
|
Thermo Scientific Acros
376871000 |
100 g | Glass bottle |
£603.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19798-81-3 | |
174.02 | |
BKLJUYPLUWUEOQ-UHFFFAOYSA-O | |
300809 | |
NC1=CC=CC(Br)=[NH+]1 |
C5H6BrN2 | |
MFCD00137843 | |
2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 | |
6-bromopyridin-2-amine |
Specifications
19798-81-3 | |
100.0 | |
Brown to Yellow | |
97.5% min. (GC) | |
MFCD00137843 | |
BKLJUYPLUWUEOQ-UHFFFAOYSA-O | |
6-bromopyridin-2-amine | |
300809 | |
98% | |
2-Amino-6-bromopyridine |
97.5 | |
88.0°C to 91.0°C | |
Authentic | |
C5H6BrN2 | |
2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663 | |
NC1=CC=CC(Br)=[NH+]1 | |
174.02 | |
173.01 | |
Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.