missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-chloropurine-9-riboside, 99%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Melting Point | 165.0°C to 175.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Specific Rotation | -35 |
Synonym | benzoic acid, 4-chlorophenyl ester, p-chlorophenol benzoate, p-chlorophenyl benzoate, benzoic acid 4-chlorophenyl ester, benzoic acid, p-chlorophenyl ester, 4-chlorophenylbenzoate, acmc-1cina, cbmicro_005244, 4-chlorophenyl benzoate |
Solubility Information | Solubility in water: .. Other solubilities: soluble in dmso,methanol |
SMILES | NC1=NC(Cl)=C2N=CN(C3OC(CO)C(O)C3O)C2=N1 |
IUPAC Name | (4-chlorophenyl) benzoate |
Show More |
Safety and Handling
Product Identifier
- 2-Amino-6-chloropurine-9-riboside
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'