missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyridine, 98%, Thermo Scientific Chemicals

Product Code. 11389815
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45644-21-1
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD00234068
InChI Key OBYJTLDIQBWBHM-UHFFFAOYSA-N
Synonym 2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine
PubChem CID 206246
IUPAC Name 6-chloropyridin-2-amine
SMILES C1=CC(=NC(=C1)Cl)N

Specifications

Melting Point 69°C to 72°C
Quantity 25 g
Beilstein 108669
Formula Weight 128.56
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chloropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'