missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific Chemicals
£40.50 - £115.00
Chemical Identifiers
CAS | 314-46-5 |
---|---|
Molecular Formula | C8H6F3NO2 |
Molecular Weight (g/mol) | 205.14 |
MDL Number | MFCD01569538 |
InChI Key | XZBQDBWIRHIZGJ-UHFFFAOYSA-N |
Synonym | 2-amino-6-trifluoromethyl benzoic acid, benzoic acid, 2-amino-6-trifluoromethyl, 6-trifluoromethyl anthranilic acid, pubchem4985, 3-amino-2-carboxybenzotrifluoride, buttpark 24\07-93, 6-trifluoromethyl anthranilicacid, 2-carboxy-3-trifluoromethyl aniline, 2-amino-6-trifluoromethyl benzoicacid |
PubChem CID | 2778508 |
IUPAC Name | 2-amino-6-(trifluoromethyl)benzoic acid |
SMILES | NC1=CC=CC(=C1C(O)=O)C(F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
314-46-5 | |
205.14 | |
XZBQDBWIRHIZGJ-UHFFFAOYSA-N | |
2778508 | |
NC1=CC=CC(=C1C(O)=O)C(F)(F)F |
C8H6F3NO2 | |
MFCD01569538 | |
2-amino-6-trifluoromethyl benzoic acid, benzoic acid, 2-amino-6-trifluoromethyl, 6-trifluoromethyl anthranilic acid, pubchem4985, 3-amino-2-carboxybenzotrifluoride, buttpark 24\07-93, 6-trifluoromethyl anthranilicacid, 2-carboxy-3-trifluoromethyl aniline, 2-amino-6-trifluoromethyl benzoicacid | |
2-amino-6-(trifluoromethyl)benzoic acid |
Specifications
314-46-5 | |
Authentic | |
Glass Bottle | |
MFCD01569538 | |
XZBQDBWIRHIZGJ-UHFFFAOYSA-N | |
2-amino-6-(trifluoromethyl)benzoic acid | |
2778508 | |
97% |
133.0°C to 136.0°C | |
96% min. (HPLC) | |
C8H6F3NO2 | |
2-amino-6-trifluoromethyl benzoic acid, benzoic acid, 2-amino-6-trifluoromethyl, 6-trifluoromethyl anthranilic acid, pubchem4985, 3-amino-2-carboxybenzotrifluoride, buttpark 24\07-93, 6-trifluoromethyl anthranilicacid, 2-carboxy-3-trifluoromethyl aniline, 2-amino-6-trifluoromethyl benzoicacid | |
NC1=CC=CC(=C1C(O)=O)C(F)(F)F | |
205.14 | |
205.14 | |
2-Amino-6-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Amino-6-(trifluoromethyl)benzoic acid, 97%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only