missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoacetophenone hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 10454765
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5468-37-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00012873
InChI Key CVXGFPPAIUELDV-UHFFFAOYSA-N
Synonym 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
PubChem CID 2723597
IUPAC Name 2-amino-1-phenylethanone;hydrochloride
SMILES Cl.NCC(=O)C1=CC=CC=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 194.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 95.0 to 105% (Argentometry)
Packaging Glass bottle
Linear Formula C6H5COCH2NH2·HCl
Quantity 1 g
Beilstein 14, 49
Merck Index 10, 7068
Formula Weight 171.63
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 2-Aminoacetophenone hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoacetophenone hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'