Learn More
2-Aminoacetophenone hydrochloride, 96%, Thermo Scientific Chemicals
£30.70 - £301.00
Chemical Identifiers
CAS | 5468-37-1 |
---|---|
Molecular Formula | C8H10ClNO |
Molecular Weight (g/mol) | 171.62 |
MDL Number | MFCD00012873 |
InChI Key | CVXGFPPAIUELDV-UHFFFAOYSA-N |
Synonym | 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride |
PubChem CID | 2723597 |
IUPAC Name | 2-amino-1-phenylethanone;hydrochloride |
SMILES | Cl.NCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10454765
|
Thermo Scientific Acros
103100010 |
1 g | Glass bottle |
£30.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10702135
|
Thermo Scientific Acros
103100050 |
5 g | Glass bottle | ||||||
10026163
|
Thermo Scientific Acros
103100250 |
25 g | Glass bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5468-37-1 | |
171.62 | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2723597 | |
Cl.NCC(=O)C1=CC=CC=C1 |
C8H10ClNO | |
MFCD00012873 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
2-amino-1-phenylethanone;hydrochloride |
Specifications
5468-37-1 | |
100.0 | |
Yellow to Beige | |
95.0 to 105% (Argentometry) | |
C6H5COCH2NH2·HCl | |
14, 49 | |
2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride | |
Cl.NCC(=O)C1=CC=CC=C1 | |
171.62 | |
171.63 | |
Crystals |
95.0 | |
194.0°C | |
Authentic | |
C8H10ClNO | |
MFCD00012873 | |
10, 7068 | |
CVXGFPPAIUELDV-UHFFFAOYSA-N | |
2-amino-1-phenylethanone;hydrochloride | |
2723597 | |
96% | |
2-Aminoacetophenone hydrochloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If eye irri
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 226-787-2
- 2-Aminoacetophenone hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only