missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ 2-Aminoacridone, Hydrochloride
Click to view available options
Quantity:
25 mg
Packaging:
1 Each
Description
2-Aminoacridone is a small, amine-containing fluorophore that is used to label reducing sugars, oligosaccharides, and glycosphingolipids. Subpicomolar detection of the derivatized products has been reported by fluorophore-assisted carbohydrate electrophoresis (FACE). Other key applications for this dye include capillary electrophoresis (CE) and high-performance liquid chromatography (HPLC)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 27918-14-5 |
Color | Green |
Packaging | 1 Each |
Molecular Formula | C13H10N2O |
Quantity | 25 mg |
Synonym | 2-aminoacridone, 2-aminoacridin-9 10h-one, 2-amino-9 10h-acridinone, 9 10h-acridinone, 2-amino, amac, 2-amino-10h-acridin-9-one, 2-amino acridone, 2-amino-9-acridone, bidd:gt0473, 2-amino-10-hydroacridin-9-one |
InChI Key | PIGCSKVALLVWKU-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 210.24 |
PubChem CID | 121965 |
ChEBI | CHEBI:31076 |
Show More |
For Research Use Only. Not for use in diagnostic procedures.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'