missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Aminobenzanilide, 95%

Product Code. 11347776
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4424-17-3
Molecular Formula C13H12N2O
Molecular Weight (g/mol) 212.25
MDL Number MFCD00034792,MFCD00017096
InChI Key FDPVTENMNDHFNK-UHFFFAOYSA-N
Synonym 2-aminobenzanilide, benzamide, 2-amino-n-phenyl, anthranilanilide, n-2-aminobenzoyl aniline, n-phenylanthranilamide, benzanilide, 2-amino, n-phenyl-2-aminobenzamide, 2-amino-n-phenyl-benzamide, 2-aminophenyl-n-benzamide, o-aminobenzanilide
PubChem CID 78142
IUPAC Name 2-amino-N-phenylbenzamide
SMILES NC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Specifications

Melting Point 130°C to 134°C
Quantity 5 g
Beilstein 782786
Formula Weight 212.25
Percent Purity 95%
Chemical Name or Material 2-Aminobenzanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'