missing translation for 'onlineSavingsMsg'
Learn More
2-Aminobenzimidazole, 97+%, Thermo Scientific Chemicals Product Code.: 11301507

2-Aminobenzimidazole, 97+%, Thermo Scientific Chemicals

Product Code. 11301507
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11301507

Brand: Thermo Scientific Alfa Aesar L02066.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzimidazole is used as receptors in chemosensor analysis. It was used in the hydrolysis of a choline carbonate. It was also used in the synthesis of imidazo[1,2-a]benzimidazoles.

Solubility
Soluble in water (Slightly).

Notes
Protect from direct light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-32-7
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005596
InChI Key JWYUFVNJZUSCSM-UHFFFAOYSA-N
Synonym 2-aminobenzimidazole, 1h-benzo d imidazol-2-amine, 2-iminobenzimidazoline, benzimidazole, 2-amino, benzimidazol-2-ylamine, 2-amino benzimidazole, caswell no. 033aa, 1h-1,3-benzodiazol-2-amine, usaf ek-4037, 2-amino-1h-benzimidazole
PubChem CID 13624
ChEBI CHEBI:27822
IUPAC Name 1H-benzimidazol-2-amine
SMILES NC1=NC2=CC=CC=C2N1

Specifications

Melting Point 228°C to 232°C
Quantity 5 g
Beilstein 116525
Solubility Information Soluble in water (Slightly).
Formula Weight 133.15
Percent Purity ≥97%
Chemical Name or Material 2-Aminobenzimidazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10214305 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberL02066.14,L02066.22,L02066.06
Lot Number10237981 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberL02066.14,L02066.22,L02066.06
Lot Number10190420 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02066.14,L02066.22,L02066.06
Lot Number10157594 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02066.14,L02066.22,L02066.06
Lot Number10214305 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL02066.14,L02066.22,L02066.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminobenzimidazole, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.