missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoethanethiol hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10424025
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
SMILES [H+].[Cl-].NCCS

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 66.0°C to 70.0°C
Color White
Density .75g/mL
Loss on Drying 1% max. (50°C, 4 hrs, 100mm Hg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NCH2CH2SH·HCl
Quantity 500 g
Beilstein 04, 286
Merck Index 15, 2776
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: very soluble in alcohol
IUPAC Name hydrogen 2-aminoethane-1-thiol chloride
Formula Weight 113.61
Percent Purity 98%
Physical Form Crystalline Powder or Pellets
Chemical Name or Material 2-Aminoethanethiol hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Aminoethanethiol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.