missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Aminoheptane, 98+%, Thermo Scientific Chemicals

Product Code. 11469907
Click to view available options
Quantity:
25 g
100 g

(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water ( 9 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N

Specifications

Density 0.768
pH 11.45
Boiling Point 142°C to 144°C
Flash Point 33°C (91°F)
Odor Amine-like
Refractive Index 1.419
Quantity 100 g
UN Number UN2733
Beilstein 635676
Sensitivity Air Sensitive
Merck Index 14,9802
Solubility Information Soluble in water ( 9g/L at 20°C).
Formula Weight 115.22
Percent Purity ≥98%
Chemical Name or Material (+/-)-2-Aminoheptane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'