missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-2-Aminoheptane sulfate, 99%, Thermo Scientific™

£67.11 - £215.07

Chemical Identifiers

CAS 6411-75-2
Molecular Formula C14H36N2O4S
Molecular Weight (g/mol) 328.512
MDL Number MFCD00050844
InChI Key XKUUMWKWUZRRPD-UHFFFAOYSA-N
Synonym sulfuric acid; bis tuaminoheptane, dsstox_cid_25845, dsstox_rid_81167, dsstox_gsid_45845, -2-aminoheptane sulfate, tuaminoheptane sulfate usp, heptan-2-amine sulfate 2:1, ^+-2-aminoheptane sulfate
PubChem CID 44134533
IUPAC Name heptan-2-amine;sulfuric acid
SMILES CCCCCC(C)N.CCCCCC(C)N.OS(=O)(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352596
View Documents Promotion Details
Alfa Aesar™
L08596.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11362596
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L08596.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6411-75-2
328.512
XKUUMWKWUZRRPD-UHFFFAOYSA-N
44134533
CCCCCC(C)N.CCCCCC(C)N.OS(=O)(=O)O
C14H36N2O4S
MFCD00050844
sulfuric acid; bis tuaminoheptane, dsstox_cid_25845, dsstox_rid_81167, dsstox_gsid_45845, -2-aminoheptane sulfate, tuaminoheptane sulfate usp, heptan-2-amine sulfate 2:1, ^+-2-aminoheptane sulfate
heptan-2-amine;sulfuric acid

Specifications

6411-75-2
C14H36N2O4S
3703311
sulfuric acid; bis tuaminoheptane, dsstox_cid_25845, dsstox_rid_81167, dsstox_gsid_45845, -2-aminoheptane sulfate, tuaminoheptane sulfate usp, heptan-2-amine sulfate 2:1, ^+-2-aminoheptane sulfate
CCCCCC(C)N.CCCCCC(C)N.OS(=O)(=O)O
328.512
328.51
(+/-)-2-Aminoheptane sulfate
289°C to 291°C
MFCD00050844
14,9802
XKUUMWKWUZRRPD-UHFFFAOYSA-N
heptan-2-amine;sulfuric acid
44134533
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 229-113-5

missing translation for 'rtecsNumber' : MQ5600000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications