missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)benzimidazole dihydrochloride hydrate, 98%

Product Code. p-7044618
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11321917

Brand: Thermo Scientific Alfa Aesar L07440.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5993-91-9 | C8H11Cl2N3 | 220.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5993-91-9
Molecular Formula C8H11Cl2N3
Molecular Weight (g/mol) 220.10
MDL Number MFCD00012677
InChI Key HAEYZZSSSUIZAN-UHFFFAOYSA-N
Synonym 1h-benzo d imidazol-2-yl methanamine dihydrochloride, 2-aminomethyl benzimidazole dihydrochloride, 1h-benzimidazole-2-methanamine, dihydrochloride, 1h-1,3-benzodiazol-2-ylmethanamine dihydrochloride, 1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 2-aminomethylbenzimidazole dihydrochloride, c-1h-benzoimidazol-2-yl-methylamine dihydrochloride, benzimidazol-2-ylmethylamine, chloride, chloride, 1-1h-1,3-benzodiazol-2-yl methanamine dihydrochloride, 1h-benzo d imidazol-2-yl methanamine 2hcl
PubChem CID 2723957
IUPAC Name 1H-benzimidazol-2-ylmethanamine;dihydrochloride
SMILES Cl.Cl.NCC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∽264°C (decomposition)
Quantity 5 g
Beilstein 3698942
Formula Weight 220.10 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-(Aminomethyl)benzimidazole dihydrochloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.