missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)indole, 97%, Thermo Scientific Chemicals

Product Code. 15470598
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21109-25-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD03422512
InChI Key RNAODKZCUVVPEN-UHFFFAOYSA-N
Synonym 1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
PubChem CID 582331
IUPAC Name 1H-indol-2-ylmethanamine
SMILES C1=CC=C2C(=C1)C=C(N2)CN

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 2-(Aminomethyl)indole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'