Learn More
2-(Aminomethyl)pyrazine, 95%, Thermo Scientific Chemicals
£96.60 - £382.00
Chemical Identifiers
CAS | 20010-99-5 |
---|---|
Molecular Formula | C5H7N3 |
Molecular Weight (g/mol) | 109.13 |
MDL Number | MFCD00673149 |
InChI Key | HQIBSDCOMQYSPF-UHFFFAOYSA-N |
Synonym | 2-aminomethylpyrazine, 2-pyrazinemethanamine, 2-aminomethyl pyrazine, pyrazin-2-yl methanamine, 1-pyrazin-2-ylmethanamine, pyrazinemethanamine, 2-aminomethyl-pyrazine, pyrazin-2-ylmethylamine, pyrazin-2-ylmethyl amine |
PubChem CID | 266781 |
IUPAC Name | pyrazin-2-ylmethanamine |
SMILES | NCC1=CN=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15254328
|
Thermo Scientific Acros
454162500 |
250 mg |
£96.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15244328
|
Thermo Scientific Acros
454160010 |
1 g |
£382.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20010-99-5 | |
109.13 | |
HQIBSDCOMQYSPF-UHFFFAOYSA-N | |
266781 | |
NCC1=CN=CC=N1 |
C5H7N3 | |
MFCD00673149 | |
2-aminomethylpyrazine, 2-pyrazinemethanamine, 2-aminomethyl pyrazine, pyrazin-2-yl methanamine, 1-pyrazin-2-ylmethanamine, pyrazinemethanamine, 2-aminomethyl-pyrazine, pyrazin-2-ylmethylamine, pyrazin-2-ylmethyl amine | |
pyrazin-2-ylmethanamine |
Specifications
20010-99-5 | |
Authentic | |
C5H7N3 | |
2-aminomethylpyrazine, 2-pyrazinemethanamine, 2-aminomethyl pyrazine, pyrazin-2-yl methanamine, 1-pyrazin-2-ylmethanamine, pyrazinemethanamine, 2-aminomethyl-pyrazine, pyrazin-2-ylmethylamine, pyrazin-2-ylmethyl amine | |
NCC1=CN=CC=N1 | |
109.13 | |
109.13 | |
2-(Aminomethyl)pyrazine |
87.0°C to 88.0°C (3.0 mmHg) | |
94% min. (GC) | |
MFCD00673149 | |
HQIBSDCOMQYSPF-UHFFFAOYSA-N | |
pyrazin-2-ylmethanamine | |
266781 | |
95% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger
- 2-(Aminomethyl)pyrazine, 95%
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only