missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenol, 99%, Thermo Scientific Chemicals

Product Code. 11404764
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

Solubility
Soluble in water (17 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O

Specifications

Melting Point 173°C to 177°C
Density 1.328
Boiling Point 150°C (11 mmHg)
Flash Point >175°C (347°F)
Odor Characteristic
Quantity 100 g
UN Number UN2512
Beilstein 606075
Merck Index 14,461
Solubility Information Soluble in water (17g/L at 20°C).
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 2-Aminophenol
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'