missing translation for 'onlineSavingsMsg'
Learn More
2-Aminopyrazine, 99%, Thermo Scientific Chemicals Product Code.: 11379679

2-Aminopyrazine, 99%, Thermo Scientific Chemicals

Product Code. 11379679
25 g
Click to view available options
Quantity:
5 g
25 g
50 g
This item is not returnable. View return policy

Product Code. 11379679

Brand: Thermo Scientific Alfa Aesar A13052.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5049-61-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD00006137
InChI Key XFTQRUTUGRCSGO-UHFFFAOYSA-N
Synonym 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazineShow MoreShow Less
PubChem CID 78747
IUPAC Name pyrazin-2-amine
SMILES C1=CN=C(C=N1)N

Specifications

Melting Point 118°C to 121°C
Quantity 25 g
Beilstein 107025
Solubility Information Soluble in water.
Formula Weight 95.11
Percent Purity 99%
Chemical Name or Material 2-Aminopyrazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS24I051 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberA13052.14,A13052.06,A13052.18
Lot Number10145757 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13052.14,A13052.06,A13052.18
Lot NumberM18H049 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13052.14,A13052.06,A13052.18
Lot Number10163815 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13052.14,A13052.06,A13052.18
Lot NumberD17S028 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13052.14,A13052.06,A13052.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminopyrazine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.