missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 10793821
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-12-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.11
MDL Number MFCD00006089
InChI Key LJXQPZWIHJMPQQ-UHFFFAOYSA-N
Synonym 2-aminopyrimidine, 2-pyrimidinamine, pyrimidin-2-ylamine, pyrimidinamine, aminopyrimidine, 2-pyrimidiylamine, pyrimidine, 2-amino, 2-pyridiylamine, 1,2-dihydro-2-iminopyrimidine, pyrimidine, amino
PubChem CID 7978
ChEBI CHEBI:38618
IUPAC Name pyrimidin-2-amine
SMILES NC1=NC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 124°C to 127°C
Color White to Yellow
Boiling Point 158°C (186.0 Mbar)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 80
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetic acid,readily soluble in alcohol,acetone and chloroform,slightly soluble in ether and benzene
Formula Weight 95.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Aminopyrimidine, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'