Learn More
2-Aminothiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
£48.80 - £140.00
Chemical Identifiers
CAS | 40283-41-8 |
---|---|
Molecular Formula | C4H4N2O2S |
Molecular Weight (g/mol) | 144.15 |
MDL Number | MFCD00859429 |
InChI Key | FCLDUALXSYSMFB-UHFFFAOYSA-N |
Synonym | 2-aminothiazole-4-carboxylic acid, 2-amino-thiazole-4-carboxylic acid, 2-amino-4-thiazolecarboxylic acid, 2-aminothiazole-4-formic acid, 4-thiazolecarboxylic acid, 2-amino, 2-aminothiazole-4-carboxylicacid, pubchem8903, acmc-209jco, 2-aminothiazole 4-carboxylic, 2-amino-4-carboxythiazole |
PubChem CID | 1501882 |
IUPAC Name | 2-amino-1,3-thiazole-4-carboxylic acid |
SMILES | C1=C(N=C(S1)N)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10748468
|
Thermo Scientific Acros
429480010 |
1 g | Glass bottle |
£48.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10412868
|
Thermo Scientific Acros
429480050 |
5 g | Glass bottle |
£140.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40283-41-8 | |
144.15 | |
FCLDUALXSYSMFB-UHFFFAOYSA-N | |
1501882 | |
C1=C(N=C(S1)N)C(=O)O |
C4H4N2O2S | |
MFCD00859429 | |
2-aminothiazole-4-carboxylic acid, 2-amino-thiazole-4-carboxylic acid, 2-amino-4-thiazolecarboxylic acid, 2-aminothiazole-4-formic acid, 4-thiazolecarboxylic acid, 2-amino, 2-aminothiazole-4-carboxylicacid, pubchem8903, acmc-209jco, 2-aminothiazole 4-carboxylic, 2-amino-4-carboxythiazole | |
2-amino-1,3-thiazole-4-carboxylic acid |
Specifications
40283-41-8 | |
100.0 | |
Colorless to Brown | |
96% min. (HPLC) | |
MFCD00859429 | |
FCLDUALXSYSMFB-UHFFFAOYSA-N | |
2-amino-1,3-thiazole-4-carboxylic acid | |
1501882 | |
97% | |
2-Aminothiazole-4-carboxylic acid |
96.0 | |
238.0°C to 239.0°C | |
Authentic | |
C4H4N2O2S | |
2-aminothiazole-4-carboxylic acid, 2-amino-thiazole-4-carboxylic acid, 2-amino-4-thiazolecarboxylic acid, 2-aminothiazole-4-formic acid, 4-thiazolecarboxylic acid, 2-amino, 2-aminothiazole-4-carboxylicacid, pubchem8903, acmc-209jco, 2-aminothiazole 4-carboxylic, 2-amino-4-carboxythiazole | |
C1=C(N=C(S1)N)C(=O)O | |
144.15 | |
144.15 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
- 2-Aminothiazole-4-carboxylic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only