missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ Two Arms Combi Light Guide With a Single Gooseneck Light Guide and a Flexible Single Light Guide

Fiber optic flex and stay light guide in a black lacquered semi-rigid gooseneck tubing and fiber optic flexible light guide in a black metal-PVC jacketing.

Brand:  SCHOTT AG Lighting and Imaging™ 120402

Product Code. 15733012

  • £247.00 / 1 unit
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Light guide with eco-friendly Puravis fiber optics.
  • Precise positioning offers complete control of light placement.
  • The combination of a gooseneck light guide 3.5 mm dia. x 500 mm and a flexible light guide 5 mm dia. x 600 mm length allows a special light scenario by using 2 different lights.
  • The single flexible light guide could be connected to the transmitted light stage 122150; to use with Cold light source KL300LED.
Specifications

Specifications

Fiber Optic Lightguide
0.4 in.
Stereo Microscopy Illumination
Gooseneck: 19.7 in., Flexible Light Guide: 23.6 in.
Puravis Glass Optical Fibers
Fiber Optic
11 mm
to Use with Cold Light Source KL300LED
Gooseneck: 500 mm, Flexible Light Guide: 600 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers