missing translation for 'onlineSavingsMsg'
Learn More
2-Azetidinone, 98%, Thermo Scientific Chemicals Product Code.: 10882451

2-Azetidinone, 98%, Thermo Scientific Chemicals

Product Code. 10882451
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10882451

Brand: Thermo Scientific Acros 432960050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-21-2
Molecular Formula C3H5NO
Molecular Weight (g/mol) 71.08
MDL Number MFCD00013328
InChI Key MNFORVFSTILPAW-UHFFFAOYSA-N
Synonym 2-azetidinone, beta-propiolactam, azetidinone, propiolactam, azetan-2-one, beta-lactam, 2-azetdinone, azetidine-2-one, acmc-209xabShow MoreShow Less
PubChem CID 136721
ChEBI CHEBI:327119
IUPAC Name azetidin-2-one
SMILES C1CNC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 72.5°C to 76.5°C
Boiling Point 106°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 71.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Azetidinone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345423 Certificate TypeCertificate of Analysis Date04/11/2024 Catalog Number432960010,432960050
Lot Number341069 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number432960010,432960050
compliance-icons
Product Identifier
  • 2-Azetidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Azetidinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.