missing translation for 'onlineSavingsMsg'
Learn More
2-Benzoylpyridine, 99+%, Thermo Scientific Chemicals Product Code.: 10152320

2-Benzoylpyridine, 99+%, Thermo Scientific Chemicals

Product Code. 10152320
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10152320

Brand: Thermo Scientific Acros 105800250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-02-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00006300
InChI Key GCSHUYKULREZSJ-UHFFFAOYSA-N
Synonym 2-benzoylpyridine, phenyl pyridin-2-yl methanone, phenyl 2-pyridyl ketone, methanone, phenyl-2-pyridinyl, pyridine, 2-benzoyl, 2-pyridyl phenyl ketone, ketone, phenyl 2-pyridyl, phenyl 2-pyridinyl methanone, phenyl-a-pyridylketone, phenyl 2-pyridyl methanone
PubChem CID 7038
IUPAC Name phenyl(pyridin-2-yl)methanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=N2

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 42.0°C to 44.0°C
Color Colorless to Yellow
Boiling Point 317.0°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C6H5CO(C5H4N)
Quantity 25 g
Beilstein 21, 330
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 183.21
Percent Purity 99+%
Physical Form Crystalline Chunks
Chemical Name or Material 2-Benzoylpyridine, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453570 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number105801000,105800250
Lot NumberA0449408 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog Number105801000,105800250
Lot NumberA0397151 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number105801000,105800250
Lot NumberA0435998 Certificate TypeCertificate of Analysis Date10/11/2021 Catalog Number105801000,105800250
Lot NumberA0404876 Certificate TypeCertificate of Analysis Date11/12/2020 Catalog Number105801000,105800250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Benzoylpyridine, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.