missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11459623
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14389-86-7
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00051940
InChI Key GMOYUTKNPLBTMT-UHFFFAOYSA-M
PubChem CID 1810581
SMILES [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 76°C to 78°C
Quantity 10 g
Synonym 2-benzyloxy benzoic acid, 2-benzyloxybenzoic acid, benzoic acid,2-phenylmethoxy, 2-phenylmethoxy benzoic acid, benzoic acid, 2-phenylmethoxy, enamine_005339, benzyloxybenzoic acid, acmc-20ao3m, 2-benzyloxy-benzoic acid, 2-benzyloxybenzoicacid
IUPAC Name 2-phenylmethoxybenzoic acid
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 2-Benzyloxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'