missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
250mg; Glass bottle
Specifications
Specifications
CAS | 1202865-21-1 |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Elemental Analysis | 74.22 to 80.41% C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Molecular Formula | C37H51O3P |
MDL Number | MFCD15143663 |
Quantity | 250mg |
Synonym | 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, 2-bis 3,5-di-tert-butyl-4-methoxyphenyl phosphanyl benzaldehyde, acmc-20ao6l, 2-bis 3,5-di-t-butyl-4-methoxyphenyl phosphino benzaldehyde, min |
Show More |
Safety and Handling
Product Identifier
- 2-(Bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphino)benzaldehyde
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'