missing translation for 'onlineSavingsMsg'
Learn More

2-(BOC-Amino)ethanethiol, 97%, Thermo Scientific Chemicals

Product Code. 10721914
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67385-09-5
Molecular Formula C7H15NO2S
Molecular Weight (g/mol) 177.26
MDL Number MFCD00274335
InChI Key GSJJCZSHYJNRPN-UHFFFAOYSA-N
Synonym 2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
PubChem CID 3017761
IUPAC Name tert-butyl N-(2-sulfanylethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCS

Specifications

Boiling Point 68.0°C (0.3 mmHg)
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3COC(O)NH(CH2)2SH
Refractive Index 1.4730 to 1.4770
Density 1.0490g/mL
Quantity 25 mL
Flash Point 109°C
Formula Weight 177.27
Percent Purity 97%
Specific Gravity 1.049
Physical Form Low Melting Solid or Viscous Liquid
Chemical Name or Material 2-(BOC-Amino)ethanethiol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(BOC-Amino)ethanethiol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'