missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(1,3-thiazol-2-yl)ethanone, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 206.06 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-1-(1, 3-thiazol-2-yl)ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'