missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2-chloropyridin-4-yl)ethanone, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Quantity | 250 mg |
Formula Weight | 234.48 |
Percent Purity | 95% |
Chemical Name or Material | 2-Bromo-1-(2-chloropyridin-4-yl)ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'