missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™
£15.00 - £253.00
Chemical Identifiers
CAS | 17570-98-8 |
---|---|
Molecular Formula | C7H7Br2NO |
Molecular Weight (g/mol) | 280.947 |
InChI Key | BYKVUGZUYJUSKD-UHFFFAOYSA-N |
Synonym | 2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374 |
PubChem CID | 2776231 |
IUPAC Name | 2-bromo-1-pyridin-2-ylethanone;hydrobromide |
SMILES | C1=CC=NC(=C1)C(=O)CBr.Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10314762
|
Thermo Scientific Maybridge
CC04005DA |
1 g | Amber Glass Bottle |
£15.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10471304
|
Thermo Scientific Maybridge
CC04005EB |
25 g | Amber Glass Bottle |
£253.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
17570-98-8 | |
280.947 | |
2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374 | |
2-bromo-1-pyridin-2-ylethanone;hydrobromide |
C7H7Br2NO | |
BYKVUGZUYJUSKD-UHFFFAOYSA-N | |
2776231 | |
C1=CC=NC(=C1)C(=O)CBr.Br |
Specifications
17570-98-8 | |
C7H7Br2NO | |
BYKVUGZUYJUSKD-UHFFFAOYSA-N | |
2-bromo-1-pyridin-2-ylethanone;hydrobromide | |
2776231 | |
90% |
90% | |
2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374 | |
C1=CC=NC(=C1)C(=O)CBr.Br | |
280.947 | |
280.95 | |
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide |