missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(3,4-dichlorophenyl)ethan-1-one, 97%, Maybridge
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 2632-10-2 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Molecular Formula | C8H5BrCl2O |
MDL Number | MFCD00051581 |
Quantity | 1 g |
Synonym | 2-bromo-1-3,4-dichlorophenyl ethanone, 3,4-dichlorophenacyl bromide, 2-bromo-3',4'-dichloroacetophenone, 2-bromo-1-3,4-dichlorophenyl ethan-1-one, 3,4-dichlorophenacylbromide, ethanone, 2-bromo-1-3,4-dichlorophenyl, 1-3,4-dichlorophenyl-2-bromoethan-1-one, pubchem3262, acmc-1coei, 3.4-dichlorophenacylbromide |
InChI Key | PAKFHEFMTRCFAU-UHFFFAOYSA-N |
SMILES | C1=CC(=C(C=C1C(=O)CBr)Cl)Cl |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'