missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene, 97%

Product Code. 11384656
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11384656

Brand: Thermo Scientific Alfa Aesar H32580.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118754-55-5 | C7H2BrCl2F3O | 309.891 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118754-55-5
Molecular Formula C7H2BrCl2F3O
Molecular Weight (g/mol) 309.891
MDL Number MFCD03412210
InChI Key CAAJZKZQEJSRJO-UHFFFAOYSA-N
Synonym 2-bromo-1,3-dichloro-5-trifluoromethoxy benzene, 2,6-dichloro-4-trifluoromethoxy bromobenzene, 4-bromo-3,5-dichloro-1-trifluoromethoxy benzene, benzene,2-bromo-1,3-dichloro-5-trifluoromethoxy, acmc-1c5si
PubChem CID 18947696
IUPAC Name 2-bromo-1,3-dichloro-5-(trifluoromethoxy)benzene
SMILES C1=C(C=C(C(=C1Cl)Br)Cl)OC(F)(F)F

Specifications

Quantity 10 g
Formula Weight 309.9
Percent Purity 97%
Chemical Name or Material 2-Bromo-1,3-dichloro-5-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.