missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 298.16 |
Percent Purity | 90% |
Chemical Name or Material | 2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'