missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(4-methyl-2-phenyl-1,3-thiazol-5-yl)-1-ethanone, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Quantity | 5 g |
Formula Weight | 296.19 |
Percent Purity | 90% |
Chemical Name or Material | 2-Bromo-1-(4-methyl-2-phenyl-1, 3-thiazol-5-yl)-1-ethanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'