missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-pentylphenyl)ethan-1-one, 95%, Thermo Scientific™

£41.40 - £206.00

Chemical Identifiers

CAS 64328-68-3
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.182
MDL Number MFCD00218837
InChI Key AVYNDJWQMUOSJZ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
PubChem CID 2775670
IUPAC Name 2-bromo-1-(4-pentylphenyl)ethanone
SMILES CCCCCC1=CC=C(C=C1)C(=O)CBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10180622
View Documents
Thermo Scientific Maybridge
BTB12841DA
1 g Amber Glass Bottle
£41.40
1g
Please to purchase this item. Need a web account? Register with us today!
10190622
View Documents
Thermo Scientific Maybridge
BTB12841EA
10 g Amber Glass Bottle
£206.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

64328-68-3
269.182
AVYNDJWQMUOSJZ-UHFFFAOYSA-N
2775670
CCCCCC1=CC=C(C=C1)C(=O)CBr
C13H17BrO
MFCD00218837
2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
2-bromo-1-(4-pentylphenyl)ethanone

Specifications

64328-68-3
100.0
C13H17BrO
MFCD00218837
AVYNDJWQMUOSJZ-UHFFFAOYSA-N
2-bromo-1-(4-pentylphenyl)ethanone
2775670
95%
94.5
95%
CH3(CH2)4C6H4C(O)CH2Br
2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
CCCCCC1=CC=C(C=C1)C(=O)CBr
269.182
269.18
2-Bromo-1-(4-pentylphenyl)ethan-1-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications