missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(trifluoromethoxy)phenyl]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10283422
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 103962-10-3
Molecular Formula C9H6BrF3O2
Molecular Weight (g/mol) 283.044
MDL Number MFCD00052333
InChI Key AOAGGWLQIILIIV-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
PubChem CID 2778691
IUPAC Name 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CF3OC6H4C(O)CH2Br
Quantity 1 g
Formula Weight 283.04
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-[4-(trifluoromethoxy)phenyl]ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'