missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(5-methyl-3-phenylisoxazol-4-yl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10486842
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10486842

Brand: Thermo Scientific Maybridge CC00505DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 104777-39-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H10BrNO2
MDL Number MFCD00173902
Quantity 1 g
Synonym 4-bromoacetyl-5-methyl-3-phenylisoxazole, 2-bromo-1-5-methyl-3-phenylisoxazol-4-yl ethan-1-one, 4-bromoacetyl-5-methyl-3-phenylisoxasole, ethanone,2-bromo-1-5-methyl-3-phenyl-4-isoxazolyl, 2-bromo-1-5-methyl-3-phenyl-1,2-oxazol-4-yl ethan-1-one, 2-bromo-1-5-methyl-3-phenyl-1,2-oxazol-4-yl ethanone, 2-bromo-1-5-methyl-3-phenylisoxazol-4-yl ethanone, acmc-1c781
InChI Key QKOOGOQWNSWJFQ-UHFFFAOYSA-N
SMILES CC1=C(C(=O)CBr)C(=NO1)C1=CC=CC=C1
IUPAC Name 2-bromo-1-(5-methyl-3-phenyl-1,2-oxazol-4-yl)ethanone
Molecular Weight (g/mol) 280.12
PubChem CID 2735531
Formula Weight 280.12
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-(5-methyl-3-phenylisoxazol-4-yl)ethan-1-one
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.