missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-indanol, 99%, Thermo Scientific Chemicals

Product Code. 11455613
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5400-80-6
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00003798
InChI Key RTESDSDXFLYAKZ-UHFFFAOYNA-N
Synonym 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783
PubChem CID 95444
IUPAC Name 2-bromo-2,3-dihydro-1H-inden-1-ol
SMILES OC1C(Br)CC2=CC=CC=C12

Specifications

Melting Point 128°C to 132°C
Quantity 250 g
Beilstein 2046877
Formula Weight 213.08
Percent Purity 99%
Chemical Name or Material 2-Bromo-1-indanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'