missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-butene, cis + trans, 98%, stab., Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.
Solubility
Not miscible or difficult to mix in water (664 mg/L 25 EST).
Notes
Store away from oxidizing agents and heat. Store away from electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.
Solubility
Not miscible or difficult to mix in water (664 mg/L 25 EST).
Notes
Store away from oxidizing agents and heat. Store away from electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
Specifications
Density | 1.328 |
Boiling Point | 82°C to 90°C |
Flash Point | 1°C (33°F) |
Odor | Recognizable |
Refractive Index | 1.461 |
Quantity | 25 g |
UN Number | UN1993 |
Beilstein | 1733329 |
Solubility Information | Not miscible or difficult to mix in water (664mg/L 25 EST). |
Formula Weight | 135 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction