missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals

Product Code. 10113020
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-19-7
Molecular Formula C4H9Br
Molecular Weight (g/mol) 137.02
MDL Number MFCD00000125
InChI Key RKSOPLXZQNSWAS-UHFFFAOYSA-N
Synonym tert-butyl bromide, t-butyl bromide, trimethylbromomethane, propane, 2-bromo-2-methyl, bromotrimethylmethane, 2-bromoisobutane, tertiarybutyl bromide, tert-butylbromide, 2-methyl-2-bromopropane, 1-bromo-1,1-dimethylethane
PubChem CID 10485
IUPAC Name 2-bromo-2-methylpropane
SMILES CC(C)(C)Br

Specifications

Melting Point -20.0°C
Density 1.1800g/mL
Boiling Point 72.0°C to 74.0°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CBr
Refractive Index 1.4269 to 1.4289
Quantity 500 mL
Beilstein 01,127
Merck Index 15,1557
Specific Gravity 1.18
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Stabilizer Potassium carbonate
Formula Weight 137.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-2-methylpropane
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'