Learn More
2-Bromo-2-nitro-1,3-propanediol, 98%, Thermo Scientific Chemicals
£27.44 - £219.09
Chemical Identifiers
CAS | 52-51-7 |
---|---|
Molecular Formula | C3H6BrNO4 |
Molecular Weight (g/mol) | 199.99 |
MDL Number | MFCD00007390 |
InChI Key | LVDKZNITIUWNER-UHFFFAOYSA-N |
Synonym | bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol |
PubChem CID | 2450 |
ChEBI | CHEBI:31306 |
IUPAC Name | 2-bromo-2-nitropropane-1,3-diol |
SMILES | C(C(CO)([N+](=O)[O-])Br)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10256060
|
Thermo Scientific Acros
158820250 |
25 g | Plastic bottle |
£27.44
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10466810
|
Thermo Scientific Acros
158821000 |
100 g | Plastic bottle |
£68.14
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10259553
|
Thermo Scientific Acros
158825000 |
500 g | Plastic bottle |
£219.09
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
52-51-7 | |
199.99 | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2450 | |
2-bromo-2-nitropropane-1,3-diol |
C3H6BrNO4 | |
MFCD00007390 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
CHEBI:31306 | |
C(C(CO)([N+](=O)[O-])Br)O |
Specifications
52-51-7 | |
100.0 | |
White to Yellow | |
97.5% min. (GC) | |
HOCH2C(Br)(NO2)CH2OH | |
01, IV, 2501 | |
bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol | |
LVDKZNITIUWNER-UHFFFAOYSA-N | |
2-bromo-2-nitropropane-1,3-diol | |
2450 | |
199.99 | |
Crystalline Powder or Crystals |
97.5 | |
>120.0°C | |
Authentic | |
C3H6BrNO4 | |
MFCD00007390 | |
15, 1457 | |
Solubility in water: 250g/L at 22°C. Other solubilities: soluble in ethanol (500g/L at 24°C),glycerol,(10g/L at 23°C),propylene glycol (400g/L at,23°C),ethyl acetate;,slightly soluble in chcl3,acetone,ether,benzene | |
C(C(CO)([N+](=O)[O-])Br)O | |
199.99 | |
CHEBI:31306 | |
98% | |
2-Bromo-2-nitro-1, 3-propanediol |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-143-
- 2-Bromo-2-nitro-1,3-propanediol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only