missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-chloropropiophenone, 98%, Thermo Scientific Chemicals

£22.31 - £79.50

Chemical Identifiers

CAS 34911-51-8
Molecular Formula C9H8BrClO
Molecular Weight (g/mol) 247.52
MDL Number MFCD03701150
InChI Key OFNMQTRHMBQQEA-UHFFFAOYNA-N
PubChem CID 4992116
IUPAC Name 2-bromo-1-(3-chlorophenyl)propan-1-one
SMILES CC(Br)C(=O)C1=CC(Cl)=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15482439
View Documents
Thermo Scientific Alfa Aesar
H64999.03
1 g
£22.31
1g
Please to purchase this item. Need a web account? Register with us today!
15492439
View Documents
Thermo Scientific Alfa Aesar
H64999.06
5 g
£79.50
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It used as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in acetonitrile, chloroform, dichloromethane and ethyl acetate.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

34911-51-8
247.52
OFNMQTRHMBQQEA-UHFFFAOYNA-N
2-bromo-1-(3-chlorophenyl)propan-1-one
C9H8BrClO
MFCD03701150
4992116
CC(Br)C(=O)C1=CC(Cl)=CC=C1

Specifications

34911-51-8
1.532 g/mL
1.577
2360404
Soluble in acetonitrile,chloroform,dichloromethane and ethyl acetate.
CC(Br)C(=O)C1=CC(Cl)=CC=C1
247.52
247.52
Liquid
Colorless to Yellow
C9H8BrClO
MFCD03701150
2-bromo-3'-chloropropiophenone, 2-bromo-1-3-chlorophenyl propan-1-one, 2-bromo-1-3-chlorophenyl-1-propanone, 3'-chloro-2-bromopropiophenone, 1-propanone,2-bromo-1-3-chlorophenyl, 1-propanone, 2-bromo-1-3-chlorophenyl, 2-bromo-1-3-chloro-phenyl-propan-1-one, bromochlorophenylpropanone, bromo-3'-chloropropiophenone, ksc573e4l
OFNMQTRHMBQQEA-UHFFFAOYNA-N
2-bromo-1-(3-chlorophenyl)propan-1-one
4992116
98%
2-Bromo-3'-chloropropiophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 252-282-1

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only