missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-methoxyacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 10605592
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-65-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.07
MDL Number MFCD00000199
InChI Key IOOHBIFQNQQUFI-UHFFFAOYSA-N
Synonym 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
PubChem CID 101294
IUPAC Name 2-bromo-1-(3-methoxyphenyl)ethanone
SMILES COC1=CC=CC(=C1)C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 64°C
Color Green-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4COCH2Br
Quantity 25 g
Formula Weight 229.07
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3'-methoxyacetophenone, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3'-methoxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'