missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-methoxyacetophenone, 98%

Product Code. 11439572
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11439572

Brand: Thermo Scientific Alfa Aesar A10716.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5000-65-7 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-65-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000199
InChI Key IOOHBIFQNQQUFI-UHFFFAOYSA-N
Synonym 2-bromo-1-3-methoxyphenyl ethanone, 2-bromo-3'-methoxyacetophenone, m-methoxyphenacyl bromide, 3'-methoxyphenacyl bromide, 3-bromoacetyl anisole, 2-bromo-1-3-methoxyphenyl ethan-1-one, 3-methoxyphenacyl bromide, bromomethyl 3-methoxyphenyl ketone, ccris 7998, 3-methoxyphenacylbromide
PubChem CID 101294
IUPAC Name 2-bromo-1-(3-methoxyphenyl)ethanone
SMILES COC1=CC=CC(=C1)C(=O)CBr

Specifications

Melting Point 60°C to 64°C
Odor Irritating
Quantity 5 g
UN Number UN3261
Beilstein 510128
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Bromo-3'-methoxyacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.