missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3-methylaniline, ≥97%, Thermo Scientific Chemicals

Product Code. 15451029
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54879-20-8
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD06411367
InChI Key VJNUZLYTGSGDHR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-3-methyl, 3-amino-2-bromotoluene, 2-bromo-3-methylbenzenamine, 2-bromo-3-methyl-benzenamine, 2-bromo-3-methylphenylamine, 2-bromo-3-methyl-phenylamine, acmc-20af7i, 2-bromo-3-methyl aniline, ksc602o1n, benzenamine,2-bromo-3-methyl
PubChem CID 603781
IUPAC Name 2-bromo-3-methylaniline
SMILES CC1=C(C(=CC=C1)N)Br

Specifications

Color Brown
Density 1.489 g/mL
Boiling Point 48°C (0.03 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.605
Quantity 5 g
UN Number UN2810
Beilstein 2079474
Formula Weight 186.05
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'