missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3-nitrobenzoic acid, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
2-Bromo-3-nitrobenzoic acid is used in preparation of 3-substituted 5-nitroisocoumarins1 and 2-bromo-3-nitrobenzaldehyde.
Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
Specifications
Specifications
Melting Point | 184°C to 186°C |
Quantity | 250 mg |
Formula Weight | 246.02 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | 2-Bromo-3-nitrobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'