missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals Product Code.: 11435540

2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11435540
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11435540

Brand: Thermo Scientific Alfa Aesar H25822.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175205-82-0
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD00153084
InChI Key OFGSIPQYQUVVPL-UHFFFAOYSA-N
Synonym 2-bromo-3-trifluoromethyl pyridine, 2-bromo-3-trifluorormethyl pyridine, pyridine, 2-bromo-3-trifluoromethyl, 2-bromo-3-trifluormethyl-pyridine, pubchem2999, acmc-1bx8n, ksc174i2d, tpc-py049, buttpark 27\06-85
PubChem CID 5193820
IUPAC Name 2-bromo-3-(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)Br)C(F)(F)F

Specifications

Melting Point 37°C to 39°C
Boiling Point 76°C (8 mmHg)
Flash Point 29°C (84°F)
Quantity 1 g
Formula Weight 226
Percent Purity 98%
Chemical Name or Material 2-Bromo-3-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-3-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.